3-Amino-4-bromo-1-methyl-1H-pyrazole 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromo-1-methyl-1H-pyrazol-3-amine, 3-Amino-4-bromo-1-methyl-1H-pyrazole, 3-Amino-4-bromo-1-methylpyrazole
OR7682-5G
APOSOR7682-5GEA
2150
CZK
APOSOR7682-5G
3-Amino-4-bromo-1-methyl-1H-pyrazole 98%
3-Amino-4-bromo-1-methyl-1H-pyrazole
Vzorec:
C₄H₆BrN₃ MW: 176,02 g/mol Bod tání: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00663065 CAS číslo: 146941-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...