Tisk…

N-[3-(Aminomethyl)phenyl]-N,N-dimethylamine 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Aminomethyl)-N,N-dimethylaniline

OR7678-1G OR7678-5G
APOSOR7678-1GEA 4300 CZK
APOSOR7678-1G APOSOR7678-5G
N-[3-(Aminomethyl)phenyl]-N,N-dimethylamine 95+%
N-[3-(Aminomethyl)phenyl]-N,N-dimethylamine
Vzorec: C₉H₁₄N₂
Bod varu: 86…89 °C/0,07mmHg
Storage Temperature: Ambient
MDL Number: MFCD00156206
CAS číslo: 57678-46-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR