Tisk…

N-(2-Aminoethyl)-N-[4-(methylsulphonyl)-2-nitrophenyl]amine 95+%

Dodavatel: Apollo Scientific

OR7675-1G OR7675-5G OR7675-25G
APOSOR7675-1GEA 5920 CZK
APOSOR7675-1G APOSOR7675-5G APOSOR7675-25G
N-(2-Aminoethyl)-N-[4-(methylsulphonyl)-2-nitrophenyl]amine 95+%
N-(2-Aminoethyl)-N-[4-(methylsulphonyl)-2-nitrophenyl]amine
Vzorec: C₉H₁₃N₃O₄S
MW: 259,29 g/mol
Bod tání: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD04037966
CAS číslo: 849035-93-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR