Tisk…

N-(4-Aminobenzyl)-N-methylamine 95+%

Dodavatel: Apollo Scientific
Nebezpečí

OR7674-1G OR7674-5G OR7674-250MG
APOSOR7674-1GEA 1120 CZK
APOSOR7674-1G APOSOR7674-5G APOSOR7674-250MG
N-(4-Aminobenzyl)-N-methylamine 95+%
N-(4-Aminobenzyl)-N-methylamine
Vzorec: C₈H₁₂N₂
Bod varu: 110 °C/0,15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00035952
CAS číslo: 38020-69-8
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR