Adamantane-1-carboxamidine hydrochloride 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR7672-1G
OR7672-250MG
APOSOR7672-1GEA
3610
CZK
APOSOR7672-1G
APOSOR7672-250MG
Adamantane-1-carboxamidine hydrochloride 95+%
Adamantane-1-carboxamidine hydrochloride
Vzorec:
C₁₁H₁₉ClN₂ Bod tání: 225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167793 CAS číslo: 50417-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...