4-{5-[(Dimethylamino)sulphonyl]-2,3-dihydro-1H-indol-1-yl}-4-oxobutanoic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR7668-1G
OR7668-5G
OR7668-25G
APOSOR7668-1GEA
5960
CZK
APOSOR7668-1G
APOSOR7668-5G
APOSOR7668-25G
4-{5-[(Dimethylamino)sulphonyl]-2,3-dihydro-1H-indol-1-yl}-4-oxobutanoic acid 95+%
4-{5-[(Dimethylamino)sulphonyl]-2,3-dihydro-1H-indol-1-yl}-4-oxobutanoic acid
Vzorec:
C₁₄H₁₈N₂O₅S MW: 326,37 g/mol Bod tání: 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00214481 CAS číslo: 393795-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...