Tisk…

2-(2-Quinoxalinyl)malondialdehyde 95+%

Dodavatel: Apollo Scientific

OR7665-25G OR7665-5G OR7665-1G
APOSOR7665-25GEA 20900 CZK
APOSOR7665-25G APOSOR7665-5G APOSOR7665-1G
2-(2-Quinoxalinyl)malondialdehyde 95+%
2-(2-Quinoxalinyl)malondialdehyde
Vzorec: C₁₁H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00216534
CAS číslo: 205744-84-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR