4-(Methylsulphonyl)-2-nitrobenzaldehyde 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR7647-1G
OR7647-250MG
APOSOR7647-1GEA
3060
CZK
APOSOR7647-1G
APOSOR7647-250MG
4-(Methylsulphonyl)-2-nitrobenzaldehyde 95+%
4-(Methylsulphonyl)-2-nitrobenzaldehyde
Vzorec:
C₈H₇NO₅S Storage Temperature: Ambient |
MDL Number:
MFCD04037929 CAS číslo: 849035-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...