2-(3-Methylphenyl)malondialdehyde 95+%
Dodavatel: Apollo Scientific
OR7646-1G
OR7646-5G
OR7646-25G
APOSOR7646-1GEA
4630
CZK
APOSOR7646-1G
APOSOR7646-5G
APOSOR7646-25G
2-(3-Methylphenyl)malondialdehyde 95+%
2-(3-Methylphenyl)malondialdehyde
Vzorec:
C₁₀H₁₀O₂ MW: 162,19 g/mol Bod tání: 81…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426050 CAS číslo: 849021-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...