2-(3,4-Dichlorophenyl)malondialdehyde 95+%
Dodavatel: Apollo Scientific
OR7636-25G
OR7636-5G
OR7636-1G
APOSOR7636-25GEA
35710
CZK
APOSOR7636-25G
APOSOR7636-5G
APOSOR7636-1G
2-(3,4-Dichlorophenyl)malondialdehyde 95+%
2-(3,4-Dichlorophenyl)malondialdehyde
Vzorec:
C₉H₆Cl₂O₂ Bod tání: 167…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD02261891 CAS číslo: 849936-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...