Tisk…

Methyl (4,6-dihydroxypyrimidin-2-ylsulphanyl)acetate

Dodavatel: Apollo Scientific

OR7630-1G OR7630-5G
APOSOR7630-1GEA 4170 CZK
APOSOR7630-1G APOSOR7630-5G
Methyl (4,6-dihydroxypyrimidin-2-ylsulphanyl)acetate
Methyl (4,6-dihydroxypyrimidin-2-ylsulphanyl)acetate
Vzorec: C₇H₈N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00489772
CAS číslo: 370077-73-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR