Tisk…

2-(4-Chloro-2-nitrophenyl)malondialdehyde 95%

Dodavatel: Apollo Scientific

OR7627-25G OR7627-5G OR7627-1G
APOSOR7627-25GEA 27280 CZK
APOSOR7627-25G APOSOR7627-5G APOSOR7627-1G
2-(4-Chloro-2-nitrophenyl)malondialdehyde 95%
2-(4-Chloro-2-nitrophenyl)malondialdehyde
Vzorec: C₉H₆ClNO₄
Bod tání: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00216498
CAS číslo: 205744-82-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR