7-Hydroxy-4-(pyridin-2-yl)coumarin 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR7615-1G
OR7615-5G
OR7615-25G
APOSOR7615-1GEA
3870
CZK
APOSOR7615-1G
APOSOR7615-5G
APOSOR7615-25G
7-Hydroxy-4-(pyridin-2-yl)coumarin 95+%
7-Hydroxy-4-(pyridin-2-yl)coumarin
Vzorec:
C₁₄H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD01941327 CAS číslo: 386704-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...