Tisk…

Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate

OR7600-5G
APOSOR7600-5GEA 2610 CZK
APOSOR7600-5G
Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate
Vzorec: C₁₀H₉BrN₂O₂
MW: 269,10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659034
CAS číslo: 67625-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR