1-Dimethylamino-2-nitroethylene 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR7588-10G
OR7588-100G
OR7588-25G
APOSOR7588-10GEA
4570
CZK
APOSOR7588-10G
APOSOR7588-100G
APOSOR7588-25G
1-Dimethylamino-2-nitroethylene 98%
1-Dimethylamino-2-nitroethylene
Vzorec:
C₄H₈N₂O₂ MW: 116,12 g/mol Bod tání: 103…105 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051519 CAS číslo: 1190-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...