Tisk…

3-Hydroxypyridine-2-carbonitrile

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Cyano-3-hydroxypyridine, 3-Hydroxypicolinonitrile

OR7552-1G OR7552-5G OR7552-25G
APOSOR7552-1GEA 622 CZK
APOSOR7552-1G APOSOR7552-5G APOSOR7552-25G
3-Hydroxypyridine-2-carbonitrile
3-Hydroxypyridine-2-carbonitrile
Vzorec: C₆H₄N₂O
MW: 120,1088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646135
CAS číslo: 932-35-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR