Tisk…

Methyl-2-(2-furyl)benzoate

Dodavatel: Apollo Scientific

OR7545-1G OR7545-250MG OR7545-500MG OR7545-5G
APOSOR7545-1GEA 7440 CZK
APOSOR7545-1G APOSOR7545-250MG APOSOR7545-500MG APOSOR7545-5G
Methyl-2-(2-furyl)benzoate
Methyl-2-(2-furyl)benzoate
Vzorec: C₁₂H₁₀O₃
MW: 202.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039126
CAS číslo: 39732-01-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR