p-Tolylhydrazinium chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Methylphenylhydrazinium chloride, 4-Tolylhydrazine hydrochloride, p-Tolylhydrazine hydrochloride, 1-(4-Methylphenyl)hydrazine hydrochloride, 4-Methylphenylhydrazine hydrochloride, 1-(4-Tolyl)hydrazine hydrochloride
OR7543-100G
OR7543-500G
APOSOR7543-100GEA
4960
CZK
APOSOR7543-100G
APOSOR7543-500G
p-Tolylhydrazinium chloride 98%
p-Tolylhydrazinium chloride
Vzorec:
C₇H₁₁ClN₂ MW: 158,63 g/mol Bod tání: 215…218 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012940 CAS číslo: 637-60-5 EINECS: 211-295-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...