Tisk…

Methyl 4-(4-formylphenyl)benzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: 4′-Formylbiphenyl-4-carboxylic acid methyl ester, Methyl 4'-formylbiphenyl-4-carboxylate, Methyl 4'-formyl-[1,1'-biphenyl]-4-carboxylate

OR7530-1G OR7530-5G
APOSOR7530-1GEA 4170 CZK
APOSOR7530-1G APOSOR7530-5G
Methyl 4-(4-formylphenyl)benzoate
Methyl 4-(4-formylphenyl)benzoate
Vzorec: C₁₅H₁₂O₃
MW: 240,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039123
CAS číslo: 70916-89-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR