Tisk…

2'-Methylbiphenyl-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2'-Methyl-[1,1'-biphenyl]-4-carboxylic acid

OR7515-1G OR7515-250MG
APOSOR7515-1GEA 2650 CZK
APOSOR7515-1G APOSOR7515-250MG
2'-Methylbiphenyl-4-carboxylic acid
2'-Methylbiphenyl-4-carboxylic acid
Vzorec: C₁₄H₁₂O₂
MW: 212,25 g/mol
Storage Temperature: Ambient
CAS číslo: 5748-43-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR