Tisk…

3'-Methoxybiphenyl-3-carbaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 3'-Methoxy-[1,1'-biphenyl]-3-carboxaldehyde, 3'-Methoxy-[1,1'-biphenyl]-3-carbaldehyde

OR7456-100MG OR7456-1G OR7456-5G
APOSOR7456-100MGEA 0 CZK
APOSOR7456-100MG APOSOR7456-1G APOSOR7456-5G
3'-Methoxybiphenyl-3-carbaldehyde
3'-Methoxybiphenyl-3-carbaldehyde
Vzorec: C₁₄H₁₂O₂
MW: 212,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424626
CAS číslo: 126485-58-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR