2-(3-Hydroxyprop-1-ynyl)benzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR7437-1G
OR7437-500MG
OR7437-250MG
APOSOR7437-1GEA
3840
CZK
APOSOR7437-1G
APOSOR7437-500MG
APOSOR7437-250MG
2-(3-Hydroxyprop-1-ynyl)benzonitrile
2-(3-Hydroxyprop-1-ynyl)benzonitrile
Vzorec:
C₁₀H₇NO Storage Temperature: Refrigerator |
MDL Number:
MFCD04039091 CAS číslo: 210884-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...