Tisk…

Coumarin-6-sulphonyl chloride 95+%

Dodavatel: Apollo Scientific
Nebezpečí

OR7425-10G OR7425-5G
APOSOR7425-10GEA 8240 CZK
APOSOR7425-10G APOSOR7425-5G
Coumarin-6-sulphonyl chloride 95+%
Coumarin-6-sulphonyl chloride
Vzorec: C₉H₅ClO₄S
Bod tání: 115…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01941320
CAS číslo: 10543-42-7
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR