Tisk…

3-(2-Furyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

OR7403-1G OR7403-250MG OR7403-50MG
APOSOR7403-1GEA 6680 CZK
APOSOR7403-1G APOSOR7403-250MG APOSOR7403-50MG
3-(2-Furyl)benzonitrile
3-(2-Furyl)benzonitrile
Vzorec: C₁₁H₇NO
Storage Temperature: Ambient
MDL Number: MFCD04039075
CAS číslo: 112598-77-3
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR