3'-Formylbiphenyl-3-carboxylic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR7387-1G
OR7387-5G
APOSOR7387-1GEA
4440
CZK
APOSOR7387-1G
APOSOR7387-5G
3'-Formylbiphenyl-3-carboxylic acid 96%
3'-Formylbiphenyl-3-carboxylic acid
Vzorec:
C₁₄H₁₀O₃ Bod tání: 174 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039068 CAS číslo: 222180-19-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...