Tisk…

2'-Formylbiphenyl-3-carboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

OR7384-1G OR7384-5G
APOSOR7384-1GEA 7440 CZK
APOSOR7384-1G APOSOR7384-5G
2'-Formylbiphenyl-3-carboxylic acid 98%
2'-Formylbiphenyl-3-carboxylic acid
Vzorec: C₁₄H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD03424616
CAS číslo: 205871-52-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR