Tisk…

4'-Chlorobiphenyl-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

OR7366-1G OR7366-5G
APOSOR7366-1GEA 787 CZK
APOSOR7366-1G APOSOR7366-5G
4'-Chlorobiphenyl-4-carboxylic acid
4'-Chlorobiphenyl-4-carboxylic acid
Vzorec: C₁₃H₉ClO₂
MW: 232,67 g/mol
Bod tání: 290…292 °C
Storage Temperature: Ambient
MDL Number: MFCD03424592
CAS číslo: 5748-41-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR