1-Boc-5-iodoindole 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Iodo-1H-indole, N-BOC protected
OR7350-1G
OR7350-5G
APOSOR7350-1GEA
2900
CZK
APOSOR7350-1G
APOSOR7350-5G
1-Boc-5-iodoindole 97%
1-Boc-5-iodoindole
Vzorec:
C₁₃H₁₄INO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09258644 CAS číslo: 374818-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...