1,1-Diphenylethylene oxide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR7342-10G
OR7342-1G
OR7342-5G
OR7342-25G
APOSOR7342-10GEA
0
CZK
APOSOR7342-10G
APOSOR7342-1G
APOSOR7342-5G
APOSOR7342-25G
1,1-Diphenylethylene oxide 97%
1,1-Diphenylethylene oxide
Vzorec:
C₁₄H₁₂O MW: 196,249 g/mol Bod tání: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040725 CAS číslo: 882-59-7 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...