Tisk…

1-(3-Nitrophenyl)-1H-pyrrole-2,5-dione

Dodavatel: Apollo Scientific

Synonyms: N-(3-nitrophenyl)maleimide

OR7302-1G OR7302-250MG
APOSOR7302-1GEA 15180 CZK
APOSOR7302-1G APOSOR7302-250MG
1-(3-Nitrophenyl)-1H-pyrrole-2,5-dione
1-(3-Nitrophenyl)-1H-pyrrole-2,5-dione
Vzorec: C₁₀H₆N₂O₄
MW: 218,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047116
CAS číslo: 7300-93-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR