Tisk…

1-(2-Methylphenyl)-1H-pyrrole-2,5-dione

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-((2-Methylphenyl)maleimide

OR7296-1G OR7296-5G
APOSOR7296-1GEA 3740 CZK
APOSOR7296-1G APOSOR7296-5G
1-(2-Methylphenyl)-1H-pyrrole-2,5-dione
1-(2-Methylphenyl)-1H-pyrrole-2,5-dione
Vzorec: C₁₁H₉NO₂
MW: 187,2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00022567
CAS číslo: 4067-01-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR