Tisk…

Methyl-3,5-dibromo-4-hydroxybenzoate 98%

Dodavatel: Apollo Scientific

OR7292-250G OR7292-50G
APOSOR7292-250GEA 14720 CZK
APOSOR7292-250G APOSOR7292-50G
Methyl-3,5-dibromo-4-hydroxybenzoate 98%
Methyl-3,5-dibromo-4-hydroxybenzoate
Vzorec: C₈H₆Br₂O₃
MW: 309,94 g/mol
Bod tání: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00016416
CAS číslo: 41727-47-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR