Tisk…

Apigenin 97%

Dodavatel: Apollo Scientific

Synonyms: 4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one

OR7265T-25G OR7265T-5G OR7265T-100G
APOSOR7265T-25GEA 3310 CZK
APOSOR7265T-25G APOSOR7265T-5G APOSOR7265T-100G
Apigenin 97%
Apigenin

A MAP kinase inhibitor. Also inhibits the proliferation of malignant tumor cells.

Vzorec: C₁₅H₁₀O₅
MW: 270,24 g/mol
Bod tání: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006831
CAS číslo: 520-36-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR