Tisk…

4-(Pentyloxy)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amyloxyphenylboronic acid, 4-Pentyloxyphenylboronic acid, (4-Pentyloxyphenyl)boronic acid, 4-Pentyloxybenzeneboronic acid, 4-n-Pentyloxybenzeneboronic acid, 4-(N-Pentyloxy)phenylboronic acid, 4-Amyloxybenzeneboronic acid

OR7247-1G OR7247-25G OR7247-5G
APOSOR7247-1GEA 622 CZK
APOSOR7247-1G APOSOR7247-25G APOSOR7247-5G
4-(Pentyloxy)phenylboronic acid
4-(Pentyloxy)phenylboronic acid
Vzorec: C₁₁H₁₇BO₃
MW: 208,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039035
CAS číslo: 146449-90-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR