Tisk…

(2-(Methoxymethyl)phenyl)boronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-methoxymethylphenyl)boronic acid, 2-(Methoxymethyl)phenylboronic acid, 2-(Methoxymethyl)benzeneboronic acid, 2-Methoxymethylphenylboronic acid

OR7230-1G OR7230-250MG OR7230-5G
APOSOR7230-1GEA 1990 CZK
APOSOR7230-1G APOSOR7230-250MG APOSOR7230-5G
(2-(Methoxymethyl)phenyl)boronic acid 98%
(2-(Methoxymethyl)phenyl)boronic acid
Vzorec: C₈H₁₁BO₃
MW: 165,98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03412110
CAS číslo: 126617-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR