5-Indoleboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid
OR7227-1G
OR7227-25G
OR7227-5G
APOSOR7227-1GEA
744
CZK
APOSOR7227-1G
APOSOR7227-25G
APOSOR7227-5G
5-Indoleboronic acid
5-Indoleboronic acid
Vzorec:
C₈H₈BNO₂ MW: 160.97 g/mol Bod tání: 170…175 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01319013 CAS číslo: 144104-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...