Tisk…

4-Indoleboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Indolylboronic acid, (1H-Indol-4-yl)boronic acid

OR7226-5G OR7226-1G
APOSOR7226-5GEA 13360 CZK
APOSOR7226-5G APOSOR7226-1G
4-Indoleboronic acid
4-Indoleboronic acid
Vzorec: C₈H₈BNO₂
MW: 160,97 g/mol
Bod tání: 184…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095175
CAS číslo: 220465-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR