Tisk…

4-(Hexyloxy)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: B-[4-(Hexyloxy)phenyl]boronic acid, 4-n-Hexyloxybenzeneboronic acid

OR7222-1G OR7222-25G OR7222-5G
APOSOR7222-1GEA 622 CZK
APOSOR7222-1G APOSOR7222-25G APOSOR7222-5G
4-(Hexyloxy)phenylboronic acid
4-(Hexyloxy)phenylboronic acid
Vzorec: C₁₂H₁₉BO₃
MW: 222,09 g/mol
Bod tání: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD04039027
CAS číslo: 121219-08-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR