3-Formyl-4-methoxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Formyl-4-methoxybenzeneboronic acid
OR7215-25G
OR7215-5G
APOSOR7215-25GEA
5390
CZK
APOSOR7215-25G
APOSOR7215-5G
3-Formyl-4-methoxyphenylboronic acid
3-Formyl-4-methoxyphenylboronic acid
Vzorec:
C₈H₉BO₄ MW: 179,97 g/mol Bod tání: 214…217 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093661 CAS číslo: 121124-97-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...