Tisk…

3,5-Dimethyl-4-methoxyphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Methoxy-3,5-dimethylphenylboronic acid, 4-Methoxy-3,5-dimethylbenzeneboronic acid

OR7208-100G OR7208-25G OR7208-5G
APOSOR7208-100GEA 20630 CZK
APOSOR7208-100G APOSOR7208-25G APOSOR7208-5G
3,5-Dimethyl-4-methoxyphenylboronic acid
3,5-Dimethyl-4-methoxyphenylboronic acid
Vzorec: C₉H₁₃BO₃
MW: 180,01 g/mol
Bod tání: 235…242 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01114648
CAS číslo: 301699-39-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR