Tisk…

3-(Cyclopropylmethoxy)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-(Cyclopropylmethoxy)phenyl)boronic acid

OR7193-1G OR7193-5G OR7193-100MG OR7193-250MG
APOSOR7193-1GEA 5720 CZK
APOSOR7193-1G APOSOR7193-5G APOSOR7193-100MG APOSOR7193-250MG
3-(Cyclopropylmethoxy)phenylboronic acid
3-(Cyclopropylmethoxy)phenylboronic acid
Vzorec: C₁₀H₁₃BO₃
MW: 192,200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039016
CAS číslo: 411229-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR