2-Bromo-5-pyridineboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid
OR7164-1G
OR7164-5G
OR7164-10G
APOSOR7164-1GEA
622
CZK
APOSOR7164-1G
APOSOR7164-5G
APOSOR7164-10G
2-Bromo-5-pyridineboronic acid 98%
2-Bromo-5-pyridineboronic acid
Vzorec:
C₅H₅BBrNO₂ MW: 201,81 g/mol Bod tání: 161…166 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD03411558 CAS číslo: 223463-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...