Tisk…
4-Benzyloxy-2-formylphenylboronic acid 97%
Dodavatel: Apollo Scientific
Synonyms: (4-(Benzyloxy)-2-formylphenyl)boronic acid
OR7158-1G OR7158-5G
APOSOR7158-1GEA 6290 CZK
APOSOR7158-1G APOSOR7158-5G
4-Benzyloxy-2-formylphenylboronic acid 97%
4-Benzyloxy-2-formylphenylboronic acid
Vzorec: C₁₄H₁₃BO₄
MW: 256,0616 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179486
CAS číslo: 139962-97-3