Tisk…

N-Benzylmaleimide

Dodavatel: Apollo Scientific
Nebezpečí

OR7122-100G OR7122-25G OR7122-5G
APOSOR7122-100GEA 3480 CZK
APOSOR7122-100G APOSOR7122-25G APOSOR7122-5G
N-Benzylmaleimide
N-Benzylmaleimide
Vzorec: C₁₁H₉NO₂
Bod tání: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00014540
CAS číslo: 1631-26-1
EINECS: 216-631-1
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR