Tisk…

(N,N-Bis-boc)-5-bromo-2-chloroaniline 97%

Dodavatel: Apollo Scientific

Synonyms: 5-Bromo-2-chloroaniline, N,N-Bis-BOC protected

OR7100-1G OR7100-250MG
APOSOR7100-1GEA 11810 CZK
APOSOR7100-1G APOSOR7100-250MG
(N,N-Bis-boc)-5-bromo-2-chloroaniline 97%
(N,N-Bis-boc)-5-bromo-2-chloroaniline
Vzorec: C₁₆H₂₁BrClNO₄
Storage Temperature: Ambient
MDL Number: MFCD09258633
CAS číslo: 929000-06-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR