2-Phenylcyclopropanecarboxylic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR7099-250MG
OR7099-1G
OR7099-5G
APOSOR7099-250MGEA
2200
CZK
APOSOR7099-250MG
APOSOR7099-1G
APOSOR7099-5G
2-Phenylcyclopropanecarboxylic acid 95+%
2-Phenylcyclopropanecarboxylic acid
Vzorec:
C₁₀H₁₀O₂ MW: 162,19 g/mol Bod tání: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066860 CAS číslo: 5685-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...