Tisk…

4-Bromo-5-methylisatin

Dodavatel: Apollo Scientific
Varování

OR7044-1G OR7044-5G OR7044-25G
APOSOR7044-1GEA 622 CZK
APOSOR7044-1G APOSOR7044-5G APOSOR7044-25G
4-Bromo-5-methylisatin
4-Bromo-5-methylisatin
Vzorec: C₉H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00560709
CAS číslo: 147149-84-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR