Bicyclo[4.1.0]heptane-7-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR7041-1G
OR7041-250MG
APOSOR7041-1GEA
8240
CZK
APOSOR7041-1G
APOSOR7041-250MG
Bicyclo[4.1.0]heptane-7-carboxylic acid
Bicyclo[4.1.0]heptane-7-carboxylic acid
Vzorec:
C₈H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00095180 CAS číslo: 41894-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...