3-Amino-2-bromo-4,6-dimethylpyridine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-4,6-dimethylpyridin-3-amine
OR7028-1G
OR7028-250MG
OR7028-500MG
OR7028-5G
APOSOR7028-1GEA
1370
CZK
APOSOR7028-1G
APOSOR7028-250MG
APOSOR7028-500MG
APOSOR7028-5G
3-Amino-2-bromo-4,6-dimethylpyridine 97%
3-Amino-2-bromo-4,6-dimethylpyridine
Vzorec:
C₇H₉BrN₂ MW: 201,0600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03731181 CAS číslo: 104829-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...