2-Amino-3-iodopyridine 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR7027-25G
OR7027-5G
OR7027-100G
APOSOR7027-25GEA
2780
CZK
APOSOR7027-25G
APOSOR7027-5G
APOSOR7027-100G
2-Amino-3-iodopyridine 99%
2-Amino-3-iodopyridine
Vzorec:
C₅H₅IN₂ MW: 220,01 g/mol Bod tání: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD01651885 CAS číslo: 104830-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...